Learn More
Cayman Chemical 4-Hydroxymephenytoin 10MG
SDP
Supplier:  Cayman Chemical 1612410 MG
Encompass_Preferred
A metabolite of (R)- and (S)-mephenytoin; formed by CYP2C19 in the liver
Catalog No. 50-297-0311