Learn More
Medchemexpress LLC 4-(Hydroxymethyl)benzeneboronic acid | 59016-93-2 | 151.96 | 25 G

Supplier: Medchemexpress LLC 25GHYW000949
4-(Hydroxymethyl)benzeneboronic acid is an important intermediate used for the synthesis of other active compounds and is involved in the synthesis of tetrahedral boronate ester. It is for research use only and serves as a laboratory chemical and for the manufacture of substances.
- Important intermediate for active compound synthesis
- Involved in the synthesis of tetrahedral boronate ester
- Suitable for laboratory chemical applications
- Used in the manufacture of substances
- Store powder at -20°C for 3 years, 4°C for 2 years
- Store in solvent at -80°C for 6 months, -20°C for 1 month
- Appearance: Solid, white to off-white
- Stable under recommended storage conditions
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.