Learn More
Medchemexpress LLC 4-(Hydroxymethyl)phenoxyacetic acid | 68858-21-9 | MFCD00057827 | ≥97.0% | 182.18 g/mol | C9H10O4 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW016413500MG
Encompass_Preferred
4-(Hydroxymethyl)phenoxyacetic acid is an acid-labile resin linkage agent (HMP linker) used in solid-phase peptide synthesis to attach peptide chains to a solid support and enable cleavage under mild acidic conditions. It is supplied for research use and commonly used in peptide assembly workflows.
  • Acid-labile linkage that enables mild cleavage from resin.
  • Designed for use in solid-phase peptide synthesis workflows.
  • Facilitates attachment of peptide chains to solid supports.
  • Compatible with standard peptide synthesis protocols.
  • Supplied in small research pack sizes for laboratory use.
Catalog No. 50-003-02195