Learn More
Medchemexpress LLC 4-(Hydroxymethyl)phenylacetic acid | 73401-74-8 | 166.18 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00412810G
Encompass_Preferred
4-(Hydroxymethyl)phenylacetic acid (4HMPAA) is a compound that functions as the primary structural component for insoluble polypeptides. It can participate in chemical reactions with DEPBT and 4-acetylaniline to generate amide intermediates. This compound is also recognized as a metabolite resulting from the breakdown of flavonoids, like quercetin, by colonic microbiota. Furthermore, 4HMPAA significantly inhibits 6-MP oxidation, demonstrating a selective inhibitory action as it does not considerably affect xanthine oxidation.
  • Functions as the main body of insoluble polypeptides.
  • Reacts with DEPBT and 4-acetylaniline to form amide intermediates.
  • A metabolite derived from the metabolism of flavonoids such as quercetin by colonic microbiota.
  • Exhibits significant inhibition of 6-MP oxidation.
  • Shows no significant inhibition on xanthine oxidation.
Catalog No. 50-004-45709