Learn More
Medchemexpress LLC 4-hydroxymethylpyrazole | 25222-43-9 | 99.7% | 98.11 | 10 G
SDP
Supplier:  Medchemexpress LLC HY3391410G
Encompass_Preferred
4-Hydroxymethylpyrazole is the primary metabolite of Fomepizole produced through hepatic oxidative metabolism. It exhibits a plasma concentration positively correlated with the administered dosage of Fomepizole and has a relatively short half-life. It also demonstrates inhibitory effects on alcohol dehydrogenase (ADH) in both humans and monkeys, though its inhibition constant is significantly higher than Fomepizole, making its in vivo impact negligible.
  • Primary metabolite of Fomepizole
  • Exhibits plasma concentration positively correlated with Fomepizole dosage
  • Relatively short half-life
  • Demonstrates inhibitory effects on alcohol dehydrogenase (ADH)
  • For research use only
Catalog No. 50-004-04143