Learn More
Medchemexpress LLC 4-Hydroxymethylpyrazole | 25222-43-9 | 99.7% | 98.11 | 100 G
SDP
Supplier:  Medchemexpress LLC HY33914100G
Encompass_Preferred
4-Hydroxymethylpyrazole is the primary metabolite of Fomepizole, produced through hepatic oxidative metabolism. It exhibits a plasma concentration positively correlated with the administered dosage of Fomepizole and has a relatively short half-life. This compound demonstrates inhibitory effects on alcohol dehydrogenase (ADH) in humans and monkeys, though its in vivo impact is negligible due to a significantly higher inhibition constant compared to Fomepizole. It is intended for research use only.
  • Primary metabolite of Fomepizole
  • Produced through hepatic oxidative metabolism
  • Exhibits plasma concentration positively correlated with administered dosage
  • Relatively short half-life
  • Demonstrates inhibitory effects on alcohol dehydrogenase (ADH)
  • For research use only
Catalog No. 50-004-03618