Learn More
Medchemexpress LLC 4-Hydroxymethylpyrazole | 25222-43-9 | 99.7% | 98.11 | 5 G
SDP
Supplier:  Medchemexpress LLC HY339145G
Encompass_Preferred
4-Hydroxymethylpyrazole is the primary metabolite of Fomepizole, produced through hepatic oxidative metabolism. It has a plasma concentration positively correlated with the administered dosage of Fomepizole and a relatively short half-life. It inhibits alcohol dehydrogenase (ADH) in humans and monkeys, but its inhibition constant is significantly higher than Fomepizole's, rendering its in vivo impact negligible.
  • Primary metabolite of Fomepizole
  • Produced through hepatic oxidative metabolism
  • Plasma concentration positively correlated with administered dosage
  • Relatively short half-life
  • Inhibits alcohol dehydrogenase (ADH) in humans and monkeys
  • In vivo impact negligible
Catalog No. 50-004-04450