Learn More
Medchemexpress LLC 4-Hydroxymethylpyrazole | 25222-43-9 | 99.74% | 98.11 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY33914
Encompass_Preferred
4-Hydroxymethylpyrazole is the primary metabolite of Fomepizole, resulting from hepatic oxidative metabolism. Its plasma concentration correlates positively with the administered Fomepizole dosage, and it has a relatively short half-life. It inhibits alcohol dehydrogenase (ADH) in humans and monkeys, although its in vivo impact is considered negligible due to a significantly higher inhibition constant than Fomepizole.
  • Functions as an endogenous metabolite
  • Demonstrates inhibitory effects on alcohol dehydrogenase
  • Suitable for research in metabolic diseases and natural products
Catalog No. 50-004-03058