Learn More
Medchemexpress LLC 4-Hydroxymethylpyrazole | 25222-43-9 | MFCD06797673 | 99.7% | 98.11 | 50 G
SDP
Supplier:  Medchemexpress LLC HY3391450G
Encompass_Preferred
4-Hydroxymethylpyrazole is the primary metabolite of Fomepizole (HY-B0876) and is produced through hepatic oxidative metabolism. It shows a plasma concentration that correlates with the administered dosage of Fomepizole and has a relatively short half-life. While it inhibits alcohol dehydrogenase (ADH) in humans and monkeys, its inhibition constant is significantly higher than that of Fomepizole, making its in vivo impact negligible.
  • Primary metabolite of Fomepizole.
  • Exhibits plasma concentration positively correlated with dosage.
  • Demonstrates inhibitory effects on alcohol dehydrogenase (ADH) in both humans and monkeys.
  • For research use only.
Catalog No. 50-004-04145