Learn More
Cayman Chemical 4-Hydroxynonal HnJ-1 M 100ug
SDP
Supplier:  Cayman Chemical 38404100 UG
Encompass_Preferred
Immunogen: 4-HNE-modified KLH • Host: Mouse • Species Reactivity: Species independent • Cross Reactivity: (+) 4-HNE-modified proteins • Applications: IF, IHC, IP, WB
Catalog No. 50-291-7092