Learn More
Cusabio Technology LLC 4-Hydroxynonenal HNE
Supplier:  Cusabio Technology LLC CSBE13412M
Encompass_Preferred
The Mouse 4-Hydroxynonenal (HNE) ELISA Kit is designed to quantitatively measure HNE levels in serum and plasma samples using a sandwich assay format This kit detects HNE with a sensitivity of 3 9 pmol/mL and a broad detection range of 15 6 pmol/mL to 1000 pmol/mL making it suitable for analyzing oxidative stress biomarkers in various experimental conditions The assay requires a sample volume of 50-100 L and delivers results within 1-5 hours Measurements are performed at an absorbance wavelength of 450 nm ensuring reliable quantification of HNE adducts formed during lipid peroxidation Ideal for research applications in metabolic disorders neurodegenerative diseases and aging studies this kit provides a standardized and efficient tool for assessing oxidative damage in mouse-derived biological samples
Catalog No. 50-000-50994