Learn More
Medchemexpress LLC 4-Hydroxyphenylacetic acid | 156-38-7 | 152.15 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYN1902R
Encompass_Preferred
4-Hydroxyphenylacetic acid is a major microbiota-derived metabolite of polyphenols involved in antioxidative action. It induces the expression of Nrf2.
  • Increases Nrf2 translocation to the nucleus and enhances the activity of phase II and antioxidant enzymes.
  • Suppresses the expression of CYP2E1.
Catalog No. 50-003-89629