Learn More
Medchemexpress LLC 4-Hydroxyphenylacetic acid | 156-38-7 | MFCD00004347 | 152.15 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN190210MM/1ML
Encompass_Preferred
4-hydroxyphenylacetic acid is a major microbiota-derived metabolite of polyphenols involved in antioxidative action. It induces the expression of Nrf2.
  • Increases Nrf2 translocation to the nucleus.
  • Enhances the activity of phase II and antioxidant enzymes.
  • Increases protein levels of nuclear Nrf2.
  • Up-regulates target genes of phase II enzymes such as UGT1A1, UGT1A9, and SULT2A1.
  • Suppresses the expression of CYP2E1.
Catalog No. 50-004-15874