Learn More
Medchemexpress LLC 4-Hydroxyphenylboronic acid pinacol ester | 269409-70-3 | 220.08 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW002879
Encompass_Preferred
This component is responsive to reactive oxygen species (ROS). It can structurally modify hyaluronic acid to create ROS-responsive intelligent nanocarriers and compound delivery systems. This modification provides hyaluronic acid-based nanocarriers with ROS responsiveness, enabling them to quickly release encapsulated substances when reactive oxygen species are present. It is applicable for research related to periodontitis and can also be utilized in the synthesis of boronic acid-modified resins.
  • Responsive to reactive oxygen species.
  • Enables structural modification of hyaluronic acid.
  • Facilitates the creation of intelligent nanocarriers and compound delivery systems.
  • Allows for rapid release of encapsulated substances in the presence of reactive oxygen species.
  • Applicable in periodontitis research.
  • Can be used for synthesizing boronic acid-modified resins.
Catalog No. 50-004-48289