Learn More
Selleck Chemical LLC 4-Hydroxyquinazoline 25mg 491-36-1 Quinazolin-4-ol, 4-Quinazolinol
SDP
Supplier:  Selleck Chemical LLC S936025MG
Encompass_Preferred
4-Hydroxyquinazoline (Quinazolin-4-ol, 4-Quinazolinol) is a PARP inhibitor with a high potency for PARP-1 with IC50 of 9.5 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1774111