Learn More
Cayman Chemical 4-HydroxyquInolIn 10g
SDP
Supplier:  Cayman Chemical 3506410 G
Encompass_Preferred
A quinoline alkaloid; is a core component of alkyl-4-hydroxyquinoline quorum sensing molecules; has been used as a precursor in the synthesis of compounds with antibacterial activity; has been used as a fluorescent detection reagent for iron(III) in bovine liver samples
Catalog No. 50-291-0472