Learn More
Medchemexpress LLC 4-hydroxyretinoic acid | 66592-72-1 | MFCD00872034 | ≥95.0% | 316.43 g/mol | C20H28O3 | 10MG

Supplier: Medchemexpress LLC HY12590410MG
4-Hydroxyretinoic acid (CAS 66592-72-1) is a naturally occurring retinoid metabolite used in biochemical and cell biology research to study retinoic acid signaling, gene regulation, and cell differentiation. It acts as a ligand for retinoic acid receptors and RXR-α and has been reported to induce apoptosis and contribute to immune regulation and neuroprotection.
- Formed from retinol and metabolized by liver enzymes.
- Ligand for retinoic acid receptors and RXR-α.
- Relevant for receptor binding, cell differentiation, and apoptosis studies.
- Molecular formula C20H28O3; molecular weight ≈ 316.4 g/mol.
- Purity ≥95.0% and solid physical form.
- Store at -80°C to maintain stability.
- Available in small research quantities, including 10 mg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.