Learn More
Medchemexpress LLC 4-hydroxyretinoic acid | 66592-72-1 | MFCD00872034 | ≥95.0% | 316.43 g·mol⁻¹ | C20H28O3 | 1MG
SDP
Supplier:  Medchemexpress LLC HY1259041MG
Encompass_Preferred
4-Hydroxyretinoic acid is a retinoid metabolite used as a research reagent to study retinoic acid receptor (RAR) and retinoid X receptor (RXR) signaling, gene expression, cell differentiation, and apoptosis. It is metabolized by liver enzymes and serves as a substrate for human UDP-glucuronosyltransferases (UGT), including UGT2B7.
  • Well-characterized retinoid for RAR/RXR studies.
  • Useful in assays of gene expression and cell differentiation.
  • Substrate for UGT-mediated metabolism and cytochrome P450 transformation.
  • Available in small quantities for research use.
  • High purity suitable for biochemical and cellular assays.
Catalog No. 50-002-35565