Learn More
Medchemexpress LLC 4'-hydroxytamoxifen | 82413-23-8 | MFCD09840778 | >98.0% | 387.51 g/mol | C26H29NO2 | 1MG
SDP
Supplier:  Medchemexpress LLC HY1244141MG
Encompass_Preferred
4'-Hydroxytamoxifen is the active metabolite of tamoxifen and acts as a selective estrogen receptor modulator (SERM). It exhibits higher affinity for the estrogen receptor than the parent drug and is commonly used in laboratory research for estrogen receptor modulation, receptor pharmacology, and CreER-inducible gene activation or knockout studies. The compound is supplied for research use with a stated purity of >98%.
  • High estrogen receptor affinity for sensitive ER studies.
  • Suitable for CreER-based inducible gene regulation experiments.
  • Available in small laboratory pack sizes for dosing and in vivo studies.
  • Research-grade material with stated purity greater than 98%.
  • Well-characterized identifiers and properties for reproducible research.
Catalog No. 50-002-38865