Learn More
Cayman Chemical 4-HydroxytryptamInfumarate 5mg
SDP
Supplier:  Cayman Chemical 90025475 MG
Encompass_Preferred
An analytical reference standard categorized as a tryptamine; an intermediate in the biosynthesis of psilocybin; intended for research and forensic applications
Catalog No. 50-289-3742