Learn More
Medchemexpress LLC 4'-hydroxyvalerophenone | 2589-71-1 | MFCD00009719 | 100.0% | 178.23 g·mol⁻1 | C11H14O2 | 50 G

Supplier: Medchemexpress LLC HYW01660450G
4'-Hydroxyvalerophenone (CAS 2589-71-1) is a small-molecule research chemical reported to inhibit 17β-hydroxysteroid dehydrogenase (IC50 ≈ 60.52 μM). It is supplied as a high-purity solid for in vitro research and analytical applications.
- High reported purity (≈99.96%).
- Molecular formula C11H14O2.
- Molecular weight 178.23 g·mol⁻1.
- Reported 17β-HSD inhibitory activity (IC50 ≈ 60.52 μM).
- Available as a 50 g solid suitable for laboratory use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.