Learn More
Medchemexpress LLC 4-ibp | 155798-08-6 | 98.9% | 420.29 g/mol | C19H21IN2O | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10015510MG
Encompass_Preferred
4-IBP is a selective σ1 receptor agonist with high affinity (Ki ≈ 1.7 nM) and moderate σ2 affinity. It is used as a research reagent in studies of sigma receptor biology and cancer models, where it has been reported to reduce cell migration and sensitize cells to pro-apoptotic agents. Supplied as a high-purity solid and as a DMSO solution for in vitro research.
  • Selective σ1 receptor agonist (Ki ≈ 1.7 nM).
  • Moderate affinity for σ2 receptor.
  • Reported to reduce cancer cell migration and sensitize cells to apoptotic agents.
  • High purity suitable for research use.
  • Available as powder and DMSO solution for in vitro studies.
  • Stable under recommended storage conditions for laboratory reagents.
Catalog No. 50-002-15738