Learn More
Selleck Chemical LLC 4-IBP
SDP
Supplier:  Selleck Chemical LLC S65615MG
Encompass_Preferred
4-IBP is a selective 1 receptor agonist with a high level of affinity for the 1 receptor (Ki 1 7 nM) and a moderate affinity for the 2 receptor (Ki 25 2 nM)
Catalog No. 50-000-00594