Learn More
Medchemexpress LLC 4-iodo-3-nitrobenzamide | 160003-66-7 | 99.7% | 100 MG

Supplier: Medchemexpress LLC HY12015100MG
Iniparib (BSI-201) is an irreversible inhibitor of poly(ADP-ribose) polymerase 1 (PARP1) used in preclinical research, notably in studies of triple-negative breast cancer. The compound is supplied as a light yellow solid with high chemical purity and recommended storage conditions to preserve stability.
- Irreversible PARP1 inhibitor for research applications.
- CAS number 160003-66-7.
- Molecular formula C7H5IN2O3.
- Molecular weight 292.03 g/mol.
- Purity about 99.7%.
- Appearance light yellow to yellow solid.
- Storage recommendations: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (2 years) or -20°C (1 year).
- Recommended DMSO stock concentrations range from 1 mM to 100 mM.
- Available in multiple solid sizes and as solution formats for flexible experimental use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.