Learn More
Medchemexpress LLC 4-Iodobenzaldehyde | 15164-44-0 | 232.02 | 10 G

Supplier: Medchemexpress LLC HYW00217910G
4-Iodobenzaldehyde is a biochemical reagent used as a biological material or organic compound for life science-related research. It is for research use only.
- Appearance: Off-white to light yellow solid
- Molecular formula: C7H5IO
- Purity: 99.86%
- Molecular weight: 232.02
- Solubility in DMSO: 100 mg/mL (431.00 mM)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.