Learn More
Matrix Scientific 4-Iodobenzonitrile, 3058-39-7, MFCD00051310, 25g
Supplier: Matrix Scientific 00669025G
Molecular Formula C7H4IN, Purity 98%, Molecular Weight 229.02, Melting Point 126-128°
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.