Learn More
Selleck Chemical LLC 4-IPP S2643-25mg
SDP
Supplier:  Selleck Chemical LLC S264325MG
Encompass_Preferred
4-IPP (4-Iodo-6-phenylpyrimidine) is an irreversible inhibitor of macrophage migration inhibitory factor (MIF) and a specific suicide substrate for MIF 4-IPP can inhibit receptor activator of NF- B ligand (RANKL)-induced osteoclastogenesis and potentiate osteoblast-mediated mineralization and bone nodule formation in vitro
Catalog No. 50-311-2877