Learn More
Medchemexpress LLC 4-Isopropylbenzoic acid | 536-66-3 | 99.9% | 164.20 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW01357110MM/1ML
Encompass_Preferred
4-Isopropylbenzoic acid, also known as Cuminic acid, is an aromatic monoterpenoid isolated from the stem bark of Bridelia retusa. It exhibits antifungal activities and acts as a reversible and uncompetitive inhibitor of mushroom tyrosinase. This compound is suitable for laboratory research, particularly in antifungal studies and enzyme inhibition investigations.
  • Shows antifungal activity for Cladosporium cladosporioides.
  • Completely inhibits the growth of P. capsici, S. sclerotiorum, and R. cerealis at 200 μg/mL.
  • Available as a solution for convenient research applications.
Catalog No. 50-004-54709