Learn More
Medchemexpress LLC 4-Isopropylbenzoic acid (Cuminic acid) | 536-66-3 | 99.9% | 164.20 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0135715G
Encompass_Preferred
4-Isopropylbenzoic acid is an aromatic monoterpenoid isolated from the stem bark of *Bridelia retusa*. It exhibits antifungal activities and is a reversible and uncompetitive inhibitor of mushroom tyrosinase.
  • Shows antifungal activity for *Cladosporium cladosporioides*.
  • Completely inhibits the growth of *P. capsici*, *S. sclerotiorum*, and *R. cerealis* at 200 μg/mL.
Catalog No. 50-004-54476