Learn More
Medchemexpress LLC 4-Isopropylbenzyl Alcohol | 536-60-7 | 10 G

Supplier: Medchemexpress LLC HYN706910G
4-Isopropylbenzyl alcohol is a chemical composition found in the essential oils from the leaves and flowers of Camellia nitidissima. It exhibits multiple biological activities, including antioxidant activity, anticancer activity, and cytotoxicity, and also inhibits the formation of advanced glycation end-products. With a molecular weight of 150.22 g/mol, this product is intended for research and development use only, not for medicinal, household, or other uses.
- Derived from Camellia nitidissima
- Exhibits antioxidant activity
- Shows anticancer activity
- Demonstrates cytotoxicity
- Inhibits advanced glycation end-products
- Colorless liquid appearance
- Melting point of 28 °C
- Boiling range of 135-136 °C at 35 hPa
- Flash point of 113 °C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.