Learn More
Medchemexpress LLC 4-Isopropylbenzyl alcohol | 536-60-7 | 100 G

Supplier: Medchemexpress LLC HYN7069100G
4-Isopropylbenzyl alcohol is a chemical compound found in the essential oils of Camellia nitidissima. This compound exhibits various biological activities, including antioxidant and anticancer properties, and has been shown to inhibit the formation of advanced glycation end-products.
- Derived from Camellia nitidissima
- Exhibits antioxidant activity
- Shows anticancer activity
- Demonstrates cytotoxicity
- Inhibits advanced glycation end-product formation
- Purity of 99.48%
- Molecular weight of 150.22
- Formula C10H14O
- Appears as a colorless to light yellow liquid
- Density of 0.982 g/cm³
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.