Learn More
Medchemexpress LLC 4-Isopropylbenzyl alcohol | 536-60-7 | 25 G

Supplier: Medchemexpress LLC 25GHYN7069
4-Isopropylbenzyl alcohol is a chemical composition found in the essential oils from the leaves and flowers of Camellia nitidissima. C. nitidissima is known for its multiple biological activities, including antioxidant activity, anticancer activity, cytotoxicity, and the ability to inhibit the formation of advanced glycation end-products. It appears as a colorless to light yellow liquid with a density of 0.982 g/cm³ and has a purity of 99.48%.
- Found in essential oils from Camellia nitidissima
- Exhibits antioxidant activity
- Shows anticancer activity
- Demonstrates cytotoxicity
- Inhibits the formation of advanced glycation end-products
- Appears as a colorless to light yellow liquid
- Density of 0.982 g/cm³
- Purity of 99.48%
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.