Learn More
Medchemexpress LLC 4-Isopropylbenzyl alcohol | 536-60-7 | 5 G

Supplier: Medchemexpress LLC HYN70695G
4-Isopropylbenzyl alcohol is a chemical component found in the essential oils derived from the leaves and flowers of Camellia nitidissima. This compound exhibits various biological activities, including antioxidant, anticancer, and cytotoxic properties, and also inhibits the formation of advanced glycation end-products. It is intended for research use only.
- Natural product originating from Camellia nitidissima
- Purity: 99.48%
- Appearance: Liquid (density: 0.982 g/cm³)
- Color: Colorless to light yellow
- Molecular weight: 150.22
- Formula: C10H14O
- Solubility: ≥ 2.5 mg/mL (16.64 mM) in various in vivo protocols
- Storage conditions:
- Pure form: -20°C for 3 years; 4°C for 2 years
- In solvent: -80°C for 6 months; -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.