Learn More
Medchemexpress LLC 4-Isopropylcatechol | 2138-43-4 | 98.0% | 152.19 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY139430100MG
Encompass_Preferred
4-Isopropylcatechol is a potent and irreversible cutaneous depigmenting agent. It significantly inhibits protein biosynthesis in mouse melanoma cells that contain high levels of tyrosinase. This compound can be utilized for research purposes to study hyperpigmentation of the skin.
  • Potent and irreversible cutaneous depigmenting agent.
  • Significantly inhibits protein biosynthesis in mouse melanoma cells containing high levels of tyrosinase.
  • Can be used to study hyperpigmentation of skins.
Catalog No. 50-003-49606