Learn More
Medchemexpress LLC 4-Isopropylcatechol | 2138-43-4 | 98.0% | 152.19 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY139430
Encompass_Preferred
4-Isopropylcatechol is a potent and irreversible cutaneous depigmenting agent. It significantly inhibits protein biosynthesis in mouse melanoma cells that contain high levels of tyrosinase. This product can be used for research on skin hyperpigmentation.
  • Potent and irreversible cutaneous depigmenting agent
  • Inhibits protein biosynthesis in mouse melanoma cells
  • Useful for research on skin hyperpigmentation
Catalog No. 50-003-48157