Learn More
Medchemexpress LLC 4-Isopropylcatechol | 2138-43-4 | 98.0% | 152.19 | 50 MG

Supplier: Medchemexpress LLC HY13943050MG
4-Isopropylcatechol is a potent and irreversible cutaneous depigmenting agent that significantly inhibits protein biosynthesis in mouse melanoma cells containing high levels of tyrosinase. It can be used to study hyperpigmentation of skins.
- Potent and irreversible cutaneous depigmenting agent
- Inhibits protein biosynthesis in mouse melanoma cells
- Useful for studying hyperpigmentation of skins
- Purity of 98.0%
- Molecular formula C9H12O2
- Appearance as an off-white to light yellow solid
- Storage at 4°C under nitrogen
- In solvent storage at -80°C for 6 months, or -20°C for 1 month, under nitrogen
- Not classified as a hazardous substance or mixture
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.