Learn More
Medchemexpress LLC 4-isothiazolecarboxamide, 3-[(4-bromo-2,6-difluorophenyl)methoxy]-5-(pyrrolidinylbutyl)amino trifluoroacetate | 2805804-54-8 | 98.0% | C22H25BrF5N5O5S | 10MG
SDP
Supplier:  Medchemexpress LLC HY13302C10MG
Encompass_Preferred
CP-547632 TFA is an orally active, ATP-competitive inhibitor of VEGFR-2 and FGF kinases with potent enzymatic activity (IC50 ≈ 11 nM for VEGFR-2 and ≈ 9 nM for FGF). Supplied as the trifluoroacetate (TFA) salt for research use, it has demonstrated antitumor efficacy in xenograft models and is characterized by comprehensive analytical documentation.
Catalog No. 50-002-58764