Learn More
Cayman Chemical 4-methoxy PCEhydrochlorIde 5mg
SDP
Supplier:  Cayman Chemical 90013565 MG
Encompass_Preferred
An analytical reference standard categorized as an arylcyclohexylamine; intended for research and forensic applications
Catalog No. 50-289-2538