Learn More
Medchemexpress LLC 4'-Methoxyagarotetrol | 123278-01-3 | 1 MG

Supplier: Medchemexpress LLC HYN81091MG
4'-Methoxyagarotetrol is a natural compound isolated from Aquilaria sinensis (Lour.). This solid, white to off-white compound has a molecular weight of 348.35 and the formula C18H20O7. It is classified as a ketone, aldehyde, and acid, and is intended for research use only.
- Natural compound
- Isolated from Aquilaria sinensis
- Molecular weight: 348.35
- Chemical formula: C18H20O7
- Appearance: solid
- Color: white to off-white
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.