Learn More
Medchemexpress LLC 4-methoxybenzenecarbothioamide | 2362-64-3 | MFCD00040993 | 95.6% | 167.23 g/mol | C8H9NOS | 25 G

Supplier: Medchemexpress LLC 25GHYW017115
4-Methoxybenzenecarbothioamide is a synthetic intermediate used in pharmaceutical and organic synthesis. Prepared by thionation of 4-methoxy benzamide, it is supplied as a light yellow to yellow solid for research and synthetic chemistry applications. Certificate of analysis indicates typical assay around 95.6% and provides handling and storage guidance.
- Synthetic intermediate for pharmaceutical and organic synthesis.
- Molecular formula C8H9NOS; molecular weight 167.23 g/mol.
- Light yellow to yellow solid suitable for synthetic use.
- Typical purity ~95.6% (per COA).
- Available in multiple pack sizes for lab-scale work.
- Store at room temperature for solid samples; follow COA guidance for solutions and long-term storage.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.