Learn More
Medchemexpress LLC 4-methoxybenzenecarbothioamide | 2362-64-3 | MFCD00040993 | 95.6% | 167.23 g·mol⁻¹ | C8H9NOS | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0171155G
Encompass_Preferred
4-Methoxybenzenecarbothioamide (CAS 2362-64-3) is a small-molecule synthetic intermediate used in medicinal chemistry and organic synthesis. Supplied as a light yellow solid (formula C8H9NOS, molecular weight 167.23 g·mol⁻¹), it is intended for research and route-development applications and has a reported purity of about 95.6%.
  • Useful building block for pharmaceutical synthesis.
  • Light yellow solid with defined physical properties.
  • Reported purity 95.6% suitable for research-grade synthesis.
  • Molecular formula C8H9NOS and molecular weight 167.23 g·mol⁻¹.
  • Stable at room temperature with recommended solvent storage conditions for extended shelf life.
  • Available in laboratory-scale pack sizes appropriate for route development and small-scale synthesis.
Catalog No. 50-003-01485