Learn More
Medchemexpress LLC 4-methoxybenzyl alcohol | 105-13-5 | MFCD00004653 | 99.9% | 138.17 g/mol | C8H10O2 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW015777100G
Encompass_Preferred
4-Methoxybenzyl alcohol is an aromatic benzyl alcohol used as a reagent and building block in organic synthesis and as an analytical standard. It is characterized by formula C8H10O2, molecular weight 138.17 g/mol, and is supplied with a high stated purity suitable for laboratory research.
  • High stated purity suitable for research and analytical use.
  • Useful building block for organic synthesis and protection/deprotection strategies.
  • Available in multiple pack sizes, including 100 g for lab-scale use.
  • Physical state varies with temperature: solid below ~22°C, liquid above ~25°C.
  • Provided with supporting documentation such as a certificate of analysis and safety data sheet.
Catalog No. 50-003-00750