Learn More
Medchemexpress LLC 4-methoxybenzyl alcohol | 105-13-5 | MFCD00004653 | 99.9% | 138.17 g/mol | C8H10O2 | 250 G
SDP
Supplier:  Medchemexpress LLC 250GHYW015777
Encompass_Preferred
4-methoxybenzyl alcohol is an aromatic primary alcohol (CAS 105-13-5) commonly used as a building block and reagent in organic synthesis and analytical chemistry. It is supplied at high purity for laboratory research and has documented physical and spectral properties to support experimental work.
  • High purity for reproducible synthetic and analytical results.
  • Useful as a benzyl-type protecting group precursor and intermediate in synthesis.
  • Compatible with common organic reactions and functional group transformations.
  • Available in multiple pack sizes to support small-scale research or larger preparations.
  • Colorless to off-white appearance with documented physical properties.
Catalog No. 50-003-02354