Learn More
Medchemexpress LLC 4-methoxybenzyl alcohol | 105-13-5 | MFCD00004653 | 99.9% | 138.17 g/mol | C8H10O2 | 500 G

Supplier: Medchemexpress LLC HYW015777500G
4-Methoxybenzyl alcohol is an organic reagent (CAS 105-13-5) commonly used in synthetic organic chemistry and research applications. It is supplied as a high-purity research chemical with specified storage recommendations and standard laboratory handling guidance.
- High purity (99.9%).
- Chemical formula C8H10O2 and molecular weight 138.17 g/mol.
- Appearance: colorless to off-white; solid below ~22°C, liquid above ~25°C.
- Suitable for research use only; not intended for human therapeutic use.
- Available in multiple pack sizes, including 500 g.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.