Learn More
Selleck Chemical LLC 4 -Methoxychalcone
SDP
Supplier:  Selleck Chemical LLC S585125MG
Encompass_Preferred
4 -Methoxychalcone found in citrus is chalcone derivative that has shown diverse pharmacological properties including anti-tumor and anti-inflammatory activities 4 -Methoxychalcone significantly enhanced adipocyte differentiation in part by its potent effects on PPAR activation and by its reverse effect on TNF-
Catalog No. 50-000-03837