Learn More
Medchemexpress LLC HY-N1387 1g Medchemexpress, 4-Methoxycinnamic acid CAS:830-09-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN13871G
Encompass_Preferred
Medchemexpress, HY-N1387 1g 4-Methoxycinnamic acid CAS:830-09-1 4-Methoxycinnamic acid is detected as natural phenylpropanoid in A. preissii. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3818
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.