Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Methoxycinnamic acid | 830-09-1 | 178.19 | 10 G
SDP

Supplier:  Medchemexpress LLC HYN138710G

Encompass_Preferred

4-Methoxycinnamic acid is a natural phenylpropanoid detected in *A. preissii*, playing a role in phenylphenalenone biosynthesis. This compound exhibits antiproliferative activity against human HepG2 and PLC-PRF-5 cells. It is supplied as an off-white to light yellow solid for research use only.

  • Natural phenylpropanoid involved in biosynthesis
  • Exhibits antiproliferative activity against human HepG2 and PLC-PRF-5 cells
  • Off-white to light yellow solid appearance
  • Soluble in DMSO at 50 mg/mL
  • Recommended storage: -20°C for powder, -80°C for in-solvent

Catalog No. 50-004-15751


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.