Learn More
Medchemexpress LLC 4-methoxyphenyl 2-(4-(3,4,5-trimethoxybenzyl)piperazin-1-yl)acetate | 113594-64-2 | MFCD00874940 | 99.0% | 430.49 | C23H30N2O6 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY4722810MG
Encompass_Preferred
KB-5492 free base is a small-molecule research compound that functions as a potent and selective sigma receptor inhibitor (IC50 = 3.15 μM) and has reported anti-ulcer activity. It is supplied as a solid (white to off-white) and as a 10 mM solution in DMSO for assay use.
  • Potent sigma receptor inhibitor (IC50 = 3.15 μM).
  • High purity, 99.0%.
  • White to off-white solid appearance.
  • Available in milligram quantities and as a DMSO solution.
  • Suitable for in vitro pharmacology and mechanism-of-action studies.
Catalog No. 50-002-98475