Learn More
Accela Chembio Inc 4-methyl-1-piperazinesulfonyl Chloride | 5g | 1688-95-5 | MFCD09731871 | 95+% | Shelf Life: 900 Days | Moisture Sensitive/store In Freezer(-20) Upon Receipt
Supplier:  Accela Chembio Inc SY0169495G
Encompass_Preferred
4-methyl-1-piperazinesulfonyl Chloride | 5g | 1688-95-5 | MFCD09731871 | 95+% | Shelf Life: 900 Days | Moisture Sensitive/store In Freezer(-20) Upon Receipt
Catalog No. 50-252-244