Learn More
Medchemexpress LLC 4-Methyl-2,3-dihydrocyclopenta[b]indol-1(4H)-one | 50776-26-6 | 99.9% | C12H11NO | 100 MG
Supplier: Medchemexpress LLC HY75357100MG
This chemical, 4-Methyl-2,3-dihydrocyclopenta[b]indol-1(4H)-one, is a light yellow to yellow solid with a molecular weight of 185.23. It functions as a drug intermediate, crucial for the synthesis of various active compounds.
- High purity (99.94%) for reliable research applications
- Key intermediate in drug synthesis
- Distinct physical appearance as a light yellow to yellow solid
- Molecular formula C12H11NO